Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKX3-2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $646.00
Specifications
Antigen | NKX3-2 |
---|---|
Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
NKX3-2 Polyclonal specifically detects NKX3-2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NKX3-2 | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
579 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:PKPRKKRSRAAFSHAQVFELERRFNHQRYLSGPERADLAASLKLTETQVKIWFQNRRYKTKRRQMAADLLASAPAAKKVAVKVLVRDDQRQYLPGEVLRPPSLLPLQPSYYYPYY | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Human | |
Bagpipe homeobox protein homolog 1, BAPX1bagpipe homeobox homolog 1 (Drosophila), Homeobox protein NK-3 homolog B, homeobox protein Nkx-3.2, MGC138171, NK3 homeobox 2, NKX3.2, NKX3BSMMD | |
NKX3-2 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title