Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nkx3.2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107193
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107193 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107193 Supplier Invitrogen™ Supplier No. PA5107193
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 83%.

This gene encodes a member of the NK family of homeobox-containing proteins. The encoded protein may play a role in skeletal development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nkx3.2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene NKX3-2
Gene Accession No. P78367
Gene Alias bagpipe homeobox gene 1 homolog; bagpipe homeobox protein homolog 1; BAPX1; homeobox protein NK-3 homolog B; Homeobox protein Nkx-3.2; homeodomain protein Nkx-3.2; NK3 homeobox 2; NKX3.2; Nkx-3.2; NKX3-2; NKX3B; SMMD
Gene Symbols NKX3-2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human NKX3-2 (aa 180-230).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 579
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.