Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKX6.1 Antibody (NKX61/2561), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279872AF594
Description
NKX6.1 Monoclonal specifically detects NKX6.1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NKX6.1 | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NKX6-1 | |
Recombinant full-length human NKX6.1 protein (Uniprot: P78426) | |
0.1 mL | |
Pancreatic Stem Cell Markers, Stem Cell Signaling Pathway, Transcription Factors and Regulators | |
Human | |
IgG2c κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
NKX61/2561 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Homeobox protein NK-6 homolog A, NK homeo box, family 6, member A, NK homeobox (Drosophila), family 6, A, NK homeobox, family 6, A, NK6 homeobox 1, NK6 transcription factor homolog A, NK6 transcription factor related, locus 1, NK6 transcription factor related, locus 1 (Drosophila), NKX6.1, NKX6Ahomeobox protein Nkx-6.1 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction