Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nkx6.1 Mouse anti-Mouse, Human, Alexa Fluor 488, Clone: R11-560, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 567608
Description
Nkx6.1 Monoclonal antibody specifically detects Nkx6.1 in Mouse, Human samples. It is validated for Intracellular staining (Flow Cytometry)
Specifications
Nkx6.1 | |
Monoclonal | |
Alexa Fluor 488 | |
Nkx6.1, NKX6-1, NKX6A, Nkx6-1, Nkx6.1, Nkx6a | |
Human Nkx6.1 Recombinant Protein | |
RUO | |
Mouse, Human | |
Liquid |
Flow Cytometry | |
R11-560 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction