Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nkx6.1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103899
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103899 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103899 Supplier Invitrogen™ Supplier No. PA5103899
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NKX61.

NKX6.1 is a bi-functional transcription factor that has effects in both the central nervous system and in the pancreas. It has both expression and repression effects on genes, leading to the proper formation of motoneurons, oligodendrocytes, and insulin-secreting beta cells. It is known to interact with Pax6 and Pdx-1, probably at or near the 60 amino acid homeobox DNA-binding domain. Due to its obvious relationship to beta cells and CNS, NKX6.1 is becoming a more interesting target for therapies in diabetes and certain brain diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nkx6.1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NKX6-1
Gene Accession No. O35762, P78426, Q99MA9
Gene Alias Drosophila NK transcription factor related, gene family 6, locus 1; homeobox protein NK-6 homolog A; Homeobox protein Nkx-6.1; homeodomain transcription factor Nkx6.1; NK homeo box, family 6, member A; NK homeobox, family 6, A; NK6 homeobox 1; NK6 transcription factor homolog A; NK6 transcription factor related locus 1; NK6 transcription factor related, locus 1; Nkx6.1; Nkx61; NKX6-1; Nkx6a
Gene Symbols NKX6-1
Host Species Rabbit
Immunogen A synthesized peptide derived from human NKX6-1(Accession P78426), corresponding to amino acid residues M209-K259.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18096, 4825, 65193
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.