Learn More
Invitrogen™ NKX6-2 Polyclonal Antibody

Description
NKX6-2 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | NKX6-2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.45 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | NKX6-2 |
| Gene Accession No. | Q9C056 |
| Gene Alias | Drosophila NK transcription factor related, gene family 6, locus 2; glial and testis specific homeobox; glial and testis-specific homeobox protein; GTX; homeobox 6B; homeobox protein NK-6 homolog B; Homeobox protein Nkx-6.2; NK homeobox family 6, B; NK6 homeobox 2; NK6 transcription factor related, locus 2; Nkx6.2; NKX6-2; NKX6B |
| Gene Symbols | NKX6-2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.