Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NKX6-2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568214
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568214 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568214 Supplier Invitrogen™ Supplier No. PA568214
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Human, Mouse, and Rat samples.

Transcription factor with repressor activity involved in the regulation of axon-glial interactions at myelin paranodes in oligodendrocytes. Binds to the consensus DNA sequence 5'-(A/T)TTAATGA-3'. In oligodendrocytes, binds to MBP and PLP1 promoter regions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKX6-2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NKX6-2
Gene Accession No. Q9C056
Gene Alias Drosophila NK transcription factor related, gene family 6, locus 2; glial and testis specific homeobox; glial and testis-specific homeobox protein; GTX; homeobox 6B; homeobox protein NK-6 homolog B; Homeobox protein Nkx-6.2; NK homeobox family 6, B; NK6 homeobox 2; NK6 transcription factor related, locus 2; Nkx6.2; NKX6-2; NKX6B
Gene Symbols NKX6-2
Host Species Rabbit
Immunogen Synthesized peptide derived from human NKX6-2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14912, 309095, 84504
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.