Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ NKX6.2, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABN1455 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN1455 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1455 Supplier MilliporeSigma™ Supplier No. ABN1455
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Homeobox protein Nkx-6.2 (UniProt Q9C056; also known as Homeobox protein NK-6 homolog B) is encoded by the NKX6-2 (also known as GTX, NKX6B) gene (Gene ID 84504) in human. Mammalian NKX gene products and their orthologs in other species are known to regulators of neuronal cell fate. Both the ventral nervous system defective (vnd) gene product in Drosophila and murine NKX proteins in spinal cord direct progenitors to adopt specific ventral neural identities. In mice, NKX6.2 is expressed in a small subset of ventral neural progenitors in a restricted spatial pattern at the border between the medial and caudal ganglionic eminence (MGE and CGE). NKX6,2 is required for specification of V1 interneurons in the spinal cord and progenitors are found to adopt a more dorsal V0 identity in the absence of NKX6,2. In addition, NKX6.2-positive brain region is shown to give rise to both parvalbumin (PV) and somatostatin (SST) cortical interneurons and in particular be enriched in a subpopulation of SST cortical interneurons that coexpress calretinin (CR).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKX6.2
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene NKX6-2, GTX, NKX6B
Gene Accession No. Q9C056
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a C-terminal region sequence of human/mouse/rat NKX6.2.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_796374
Test Specificity This rabbit polyclonal antibody detected the exogenously expressed human NKX6.2 in transfected HEK293T cells, while no immunoreactive bands were detected in lysates from untransfected cells. The immunogen sequence lies within the region C-terminal to the Homeobox and is 100% conserved among human, mouse, and rat species.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.