Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ NKY 80
GREENER_CHOICESDP

Catalog No. 507150R Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
507150R 50 mg
507110R 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 507150R Supplier R&D Systems™ Supplier No. 5071/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Adenylyl cyclase inhibitor

Inhibitor of adenylyl cyclase (AC). Exhibits greater affinity for AC5 over AC3 and AC2 (IC50 values are 8.3 μM, 132 μM and 1.7 mM respectively).

Chemical Identifiers

CAS 299442-43-6
Molecular Formula C12H11N3O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 229.23
Physical Form Solid
Color White
Purity >98%
Molecular Weight (g/mol) 229.24
Chemical Name or Material NKY 80
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.