Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NLE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106694
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106694 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106694 Supplier Invitrogen™ Supplier No. PA5106694
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NLE1.

NLE1 plays a role in regulating Notch activity, in regulating the expression of CDKN1A and several members of the Wnt pathway, probably via its effects on Notch activity. NLE1 is required during embryogenesis for inner mass cell survival.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLE1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NLE1
Gene Accession No. Q9NVX2
Gene Alias AL022765; BC018399; FLJ10458; HUSSY-07; l11Jus1; l11Jus4; Nle; Nle1; Notchless gene homolog; notchless homolog 1; notchless homolog 1 (Drosophila); notchless protein homolog 1; novel WD40 repeat containing protein; RGD1308301
Gene Symbols NLE1
Host Species Rabbit
Immunogen A synthesized peptide derived from human NLE1(Accession Q9NVX2), corresponding to amino acid residues A300-D350.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54475
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.