Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NLE1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550075

Catalog No. PA550075


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total NLE1 protein.

NLE1 plays a role in regulating Notch activity, in regulating the expression of CDKN1A and several members of the Wnt pathway, probably via its effects on Notch activity. NLE1 is required during embryogenesis for inner mass cell survival.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NLE1
Polyclonal
Unconjugated
NLE1
AL022765; BC018399; FLJ10458; HUSSY-07; l11Jus1; l11Jus4; Nle; Nle1; Notchless gene homolog; notchless homolog 1; notchless homolog 1 (Drosophila); notchless protein homolog 1; novel WD40 repeat containing protein; RGD1308301
Rabbit
Antigen affinity chromatography
RUO
217011, 303372, 54475
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8VEJ4, Q9NVX2
NLE1
Synthesized peptide derived from internal of human NLE1.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.