Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NLGN3 Monoclonal Antibody (N110/29), APC
GREENER_CHOICE

Catalog No. PIMA545697
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545697 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545697 Supplier Invitrogen™ Supplier No. MA545697
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse: 99% identity (118/119 amino acids identical). Human: 98% identity (116/119 amino acids identical) ~60% identity with Neuroligin-1. ~40% identity with Neuroligin-2. 1 μg/mL of MA5-45697 was sufficient for detection of Neuroligin 3 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 110kDa. Does not cross-react with Neuroligin-1, -2, -4 or -4. This antibody was formerly sold as clone S110-29.

This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses. Mutations in this gene may be associated with autism and Asperger syndrome. Multiple transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLGN3
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N110/29
Concentration 1 mg/mL
Conjugate APC
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene NLGN3
Gene Accession No. Q62889, Q8BYM5, Q9NZ94
Gene Alias A230085M13Rik; ASPGX1; AUTSX1; Gliotactin homolog; HNL3; KIAA1480; neuroligin 3; neuroligin 3 protein; neuroligin-3; NL3; NLGN3
Gene Symbols NLGN3
Host Species Mouse
Immunogen Fusion protein amino acids 730-848 (intracellular C-terminus) of rat Neuroligin-3.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171297, 245537, 54413
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.