Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nlgn4l, Mouse anti-Mouse, ATTO594, Clone: S98-7, Abnova™

Catalog No. 89338595 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-338-595 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-338-595 Supplier Abnova Corporation Supplier No. MAB18670
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen Nlgn4l
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone S98-7
Conjugate Atto 594
Dilution Immunocytochemistry Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide).
Gene neuroligin 4-like
Gene Alias NL-4/NL4/Nlgn4/Nlgn4x
Gene Symbols Nlgn4l
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 782-945 at C-terminus of mouse Nlgn4l.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100113365
Target Species Mouse
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.