Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NLGN4X (Neuroligin-4) Monoclonal Antibody (CSTEM30), eBioscience™, Invitrogen™

Catalog No. 14915837
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-915-837 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-915-837 Supplier Invitrogen™ Supplier No. 14915837
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This monoclonal antibody CSTEM30 reacts with human Neuroligin-4, X linked, also known as NLGN4X, NLGN4, Neuroligin-4, and Neuroligin 4X. Applications Reported: This CSTEM30 antibody has been reported for use in immunocytochemical staining and flow cytometric analysis. (Fluorochrome-conjugated CSTEM30 is recommended for use in flow cytometery.) Applications Tested: This CSTEM30 antibody has been tested by immunocytochemistry on formaldehyde-fixed and permeabilized human iPSCs, and by flow cytometry on human iPSCs. For immunocytochemistry, this can be used at less than or equal to 5 μg/mL. For flow cytometry, this antibody can be used at less than or equal to 0.5 μg/test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

Neuroligin-4, X linked (also known as NLGN4X, NLGN4, Neuroligin-4, and Neuroligin 4X) is a postsynaptic cell-adhesion molecule that interacts with presynaptic cell-surface neurexins. These interactions bridge together the synaptic cleft and have an essential role in synaptic transmissions between neurons. There are 5 neuroligin genes in humans: NLGN1, NLGN2, NLGN3, NLGN4X, and NLGN4Y. Mutations in NLGN3 and NLGN4X are linked to Autism Spectrum Disorders (ASD). The expression of neuroligin-4, X-linked has been reported on human pluripotent stem cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLGN4X (Neuroligin-4)
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone CSTEM30
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.09% sodium azide; pH 7.2
Gene NLGN4X
Gene Accession No. Q8N0W4
Gene Alias ASPGX2; AUTSX2; HLNX; HNL4X; HNLX; KIAA1260; MGC22376; neuroligin 4, X-linked; neuroligin 4-like; Neuroligin X; Neuroligin-4, X-linked; NL4; NL-4; NLGN; Nlgn4; Nlgn4l; NLGN4X; UNQ365/PRO701
Gene Symbols NLGN4X
Host Species Mouse
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57502
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.