Learn More
Invitrogen™ NLGN4X (Neuroligin-4) Monoclonal Antibody (N98/7), APC
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA545693
Catalog No. PIMA545693
Only null left
Add to Cart
Description
Description
~45% identity with Neuroligin-1, -2 and -3. 1 μg/mL of MA5-45693 was sufficient for detection of Neuroligin 4 in 20 μg of mouse brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 125-130kDa. Does not cross-react with other Neuroligins. This antibody was formerly sold as clone S98-7.
Neuroligin-4, X linked (also known as NLGN4X, NLGN4, Neuroligin-4, and Neuroligin 4X) is a postsynaptic cell-adhesion molecule that interacts with presynaptic cell-surface neurexins. These interactions bridge together the synaptic cleft and have an essential role in synaptic transmissions between neurons. There are 5 neuroligin genes in humans: NLGN1, NLGN2, NLGN3, NLGN4X, and NLGN4Y. Mutations in NLGN3 and NLGN4X are linked to Autism Spectrum Disorders (ASD). The expression of neuroligin-4, X-linked has been reported on human pluripotent stem cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NLGN4X (Neuroligin-4)
Monoclonal
1 mg/mL
2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
B0F2B4
Nlgn4l
Fusion protein amino acids 782-945 (intracellular C-terminus) of mouse Neuroligin 4.
100 μg
Primary
Mouse, Rat
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
N98/7
APC
Nlgn4l
ASPGX2; AUTSX2; HLNX; HNL4X; HNLX; KIAA1260; MGC22376; neuroligin 4, X-linked; neuroligin 4-like; Neuroligin X; Neuroligin-4, X-linked; NL4; NL-4; NLGN; Nlgn4; Nlgn4l; NLGN4X; UNQ365/PRO701
Mouse
Protein G
RUO
100113365
4°C
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.