Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NLGN4X (Neuroligin-4) Monoclonal Antibody (N98/7), FITC
GREENER_CHOICE

Catalog No. PIMA545694
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545694 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545694 Supplier Invitrogen™ Supplier No. MA545694
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

~45% identity with Neuroligin-1, -2 and -3. 1 μg/mL of MA5-45694 was sufficient for detection of Neuroligin 4 in 20 μg of mouse brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 125-130kDa. Does not cross-react with other Neuroligins. This antibody was formerly sold as clone S98-7.

Neuroligin-4, X linked (also known as NLGN4X, NLGN4, Neuroligin-4, and Neuroligin 4X) is a postsynaptic cell-adhesion molecule that interacts with presynaptic cell-surface neurexins. These interactions bridge together the synaptic cleft and have an essential role in synaptic transmissions between neurons. There are 5 neuroligin genes in humans: NLGN1, NLGN2, NLGN3, NLGN4X, and NLGN4Y. Mutations in NLGN3 and NLGN4X are linked to Autism Spectrum Disorders (ASD). The expression of neuroligin-4, X-linked has been reported on human pluripotent stem cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLGN4X (Neuroligin-4)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N98/7
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Gene Nlgn4l
Gene Accession No. B0F2B4
Gene Alias ASPGX2; AUTSX2; HLNX; HNL4X; HNLX; KIAA1260; MGC22376; neuroligin 4, X-linked; neuroligin 4-like; Neuroligin X; Neuroligin-4, X-linked; NL4; NL-4; NLGN; Nlgn4; Nlgn4l; NLGN4X; UNQ365/PRO701
Gene Symbols Nlgn4l
Host Species Mouse
Immunogen Fusion protein amino acids 782-945 (intracellular C-terminus) of mouse Neuroligin 4.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100113365
Target Species Mouse, Rat
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.