Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NLGN4X Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547703
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547703 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547703 Supplier Invitrogen™ Supplier No. PA547703
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, less than 5% cross-reactivity with recombinant rat NLGN1, recombinant human (rh) NLGN2 and rhNLGN3 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

Neuroligin-4, X linked (also known as NLGN4X, NLGN4, Neuroligin-4, and Neuroligin 4X) is a postsynaptic cell-adhesion molecule that interacts with presynaptic cell-surface neurexins. These interactions bridge together the synaptic cleft and have an essential role in synaptic transmissions between neurons. There are 5 neuroligin genes in humans: NLGN1, NLGN2, NLGN3, NLGN4X, and NLGN4Y. Mutations in NLGN3 and NLGN4X are linked to Autism Spectrum Disorders (ASD). The expression of neuroligin-4, X-linked has been reported on human pluripotent stem cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLGN4X
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene NLGN4X
Gene Accession No. Q8N0W4
Gene Alias ASPGX2; AUTSX2; HLNX; HNL4X; HNLX; KIAA1260; MGC22376; neuroligin 4, X-linked; neuroligin 4-like; Neuroligin X; Neuroligin-4, X-linked; NL4; NL-4; NLGN; Nlgn4; Nlgn4l; NLGN4X; UNQ365/PRO701
Gene Symbols NLGN4X
Host Species Sheep
Immunogen E. coli-derived recombinant human Neuroligin 4/NLGN4 Met618-Thr673.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57502
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.