Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NLGN4X Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110032
Description
Immunogen sequence: YYKKDKRRHE THRRPSPQRN TTNDIAHIQN EEIMSLQMKQ LEHDHECESL QAHDTLRLTC PPDYTLTLRR SPDDIPLMTP NTITMIPNTL TGMQPLHTFN TFSGGQNSTN LPHGHSTTRV.
Neuroligin-4, X linked (also known as NLGN4X, NLGN4, Neuroligin-4, and Neuroligin 4X) is a postsynaptic cell-adhesion molecule that interacts with presynaptic cell-surface neurexins. These interactions bridge together the synaptic cleft and have an essential role in synaptic transmissions between neurons. There are 5 neuroligin genes in humans: NLGN1, NLGN2, NLGN3, NLGN4X, and NLGN4Y. Mutations in NLGN3 and NLGN4X are linked to Autism Spectrum Disorders (ASD). The expression of neuroligin-4, X-linked has been reported on human pluripotent stem cells.
Specifications
| NLGN4X | |
| Polyclonal | |
| Unconjugated | |
| NLGN4X | |
| ASPGX2; AUTSX2; HLNX; HNL4X; HNLX; KIAA1260; MGC22376; neuroligin 4, X-linked; neuroligin 4-like; Neuroligin X; Neuroligin-4, X-linked; NL4; NL-4; NLGN; Nlgn4; Nlgn4l; NLGN4X; UNQ365/PRO701 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 100113365, 57502 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.58 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B0F2B4, Q8N0W4 | |
| Nlgn4l, NLGN4X | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 697-816 of human NLGN4X (NP_065793.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction