Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NLGN4Y Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576169
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576169 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576169 Supplier Invitrogen™ Supplier No. PA576169
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Neuroligins, such as NLGN4Y, are cell adhesion molecules present at the postsynaptic side of the synapse and may be essential for the formation of functional synapses (Jamain et al., 2003 [PubMed 12669065]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLGN4Y
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene NLGN4Y
Gene Accession No. Q8NFZ3
Gene Alias ASPGX2; AUTSX2; HLNX; HNL4X; HNLX; KIAA0951; KIAA1260; neuroligin 4, X-linked; neuroligin 4, Y-linked; Neuroligin X; Neuroligin Y; Neuroligin-4, X-linked; neuroligin-4, Y-linked; NLGN4; NLGN4X; NLGN4Y; UNQ365/PRO701
Gene Symbols NLGN4Y
Host Species Rabbit
Immunogen Recombinant protein of human NLGN4Y.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22829
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.