Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NLGN4Y Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113136
Description
NLGN4Y Polyclonal Antibody for Western Blot, IHC (P), ELISA
Neuroligins, such as NLGN4Y, are cell adhesion molecules present at the postsynaptic side of the synapse and may be essential for the formation of functional synapses (Jamain et al., 2003 [PubMed 12669065]).
Specifications
NLGN4Y | |
Polyclonal | |
Unconjugated | |
NLGN4Y | |
ASPGX2; AUTSX2; HLNX; HNL4X; HNLX; KIAA0951; KIAA1260; neuroligin 4, X-linked; neuroligin 4, Y-linked; Neuroligin X; Neuroligin Y; Neuroligin-4, X-linked; neuroligin-4, Y-linked; NLGN4; NLGN4X; NLGN4Y; UNQ365/PRO701 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22829 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.86 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8NFZ3 | |
NLGN4Y | |
Recombinant Human Neuroligin-4, Y-linked protein (682-816AA). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction