Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NLGN4Y Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120888
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120888 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120888 Supplier Invitrogen™ Supplier No. PA5120888
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-87MG, U-251MG, HeLa, A-549, Mouse brain, Mouse testis, Rat brain, Rat testis. The target is usually found in the following locations: Cell junction, Cell membrane, Single-pass type I membrane protein, postsynaptic cell membrane, postsynaptic density, synapse. Immunogen sequence: DMLPIWFTTS LDTLMTYVQD QNEDCLYLNI YVPMEDGTNI KRNADDITSN DHGEDKDIHE QNSKKPVMVY IHGGSYMEGT GNMIDGSILA SYGNVIVITI NYRLGILGMQ EARLCGSSKM FNYFKSPFTN LINFF.

Neuroligins, such as NLGN4Y, are cell adhesion molecules present at the postsynaptic side of the synapse and may be essential for the formation of functional synapses (Jamain et al., 2003 [PubMed 12669065]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLGN4Y
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NLGN4Y
Gene Accession No. Q8NFZ3
Gene Alias ASPGX2; AUTSX2; HLNX; HNL4X; HNLX; KIAA0951; KIAA1260; neuroligin 4, X-linked; neuroligin 4, Y-linked; Neuroligin X; Neuroligin Y; Neuroligin-4, X-linked; neuroligin-4, Y-linked; NLGN4; NLGN4X; NLGN4Y; UNQ365/PRO701
Gene Symbols NLGN4Y
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 122-256 of human NLGN4Y (NP_001157710.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22829
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.