Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NLK Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299830PCP
Description
NLK Polyclonal antibody specifically detects NLK in Human samples. It is validated for Western Blot, ELISASpecifications
NLK | |
Polyclonal | |
Western Blot, ELISA | |
DKFZp761G1211, EC 2.7.11, EC 2.7.11.24, FLJ21033, LAK1, nemo like kinase, nemo-like kinaseserine/threonine-protein kinase NLK, Protein LAK1 | |
Produced in rabbits immunized with purified, recombinant Human NLK (Uniprot#: Q9UBE8; Val121-Glu527) | |
0.1 mL | |
Protein Kinase, Wnt Signaling Pathway | |
51701 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction