Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NLK Antibody (SR2028), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | NLK |
---|---|
Clone | SR2028 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50 |
Applications | Western Blot, Flow Cytometry |
Classification | Monoclonal |
Description
NLK Monoclonal antibody specifically detects NLK in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
NLK | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
DKFZp761G1211, EC 2.7.11, EC 2.7.11.24, FLJ21033, LAK1, nemo like kinase, nemo-like kinaseserine/threonine-protein kinase NLK, Protein LAK1 | |
A synthesized peptide derived from human NLK (Uniprot #: Q9UBE8) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2028 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Protein Kinase, Wnt Signaling Pathway | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
51701 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title