Learn More
Invitrogen™ NLK Polyclonal Antibody

Description
Immunogen sequence: AAATVKAHHH QHSHHPQQQL DIEPDRPIGY GAFGVVWSVT DPRDGKRVAL KKMPNVFQNL VSCK Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
Specifications
Specifications
| Antigen | NLK |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | NLK |
| Gene Accession No. | Q9UBE8 |
| Gene Alias | AI194375; LAK1; nemo like kinase; nemo-like kinase; NLK; Protein LAK1; RGD1561602; serine/threonine protein kinase NLK; serine/threonine-protein kinase NLK |
| Gene Symbols | NLK |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.