Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NLN Monoclonal Antibody (OTI1D6)
GREENER_CHOICE

Catalog No. PIMA525842
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525842 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525842 Supplier Invitrogen™ Supplier No. MA525842
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NLN Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

This gene encodes a member of the metallopeptidase M3 protein family that cleaves neurotensin at the Pro10-Tyr11 bond, leading to the formation of neurotensin(1-10) and neurotensin(11-13). The encoded protein is likely involved in the termination of the neurotensinergic signal in the central nervous system and in the gastrointestinal tract.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLN
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1D6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Nln
Gene Accession No. P42676, Q91YP2, Q9BYT8
Gene Alias 4930472G13Rik; AGTBP; angiotensin binding protein; angiotensin-binding protein; C79345; endopeptidase 24.16; EP24.16; KIAA1226; MEP; microsomal endopeptidase; Mitochondrial oligopeptidase M; MOP; neurolysin; neurolysin (metallopeptidase M3 family); neurolysin, mitochondrial; Neurotensin endopeptidase; NLN
Gene Symbols Nln
Host Species Mouse
Immunogen Full length human recombinant protein of NLN produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117041, 57486, 75805
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.