Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLN Monoclonal Antibody (OTI3C2), TrueMAB™, OriGene
SDP

Catalog No. 502540555
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0555 30 μL
50-167-4794 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0555 Supplier OriGene Supplier No. TA504021S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NLN Monoclonal antibody specifically detects NLN in Canine, Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen NLN
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI3C2
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DPP9
Gene Accession No. Q86TI2
Gene Alias 4930472G13Rik; AGTBP; angiotensin binding protein; angiotensin-binding protein; C79345; endopeptidase 24.16; EP24.16; KIAA1226; MEP; microsomal endopeptidase; Mitochondrial oligopeptidase M; MOP; neurolysin; neurolysin (metallopeptidase M3 family); neurolysin, mitochondrial; Neurotensin endopeptidase; NLN
Host Species Mouse
Immunogen Full length human recombinant protein of human DPP9 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 485033, 91039
Target Species Canine, Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.