Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NLN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588992
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588992 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588992 Supplier Invitrogen™ Supplier No. PA588992
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-87MG, A-549, Rat liver; Cellular Location: Cytoplasm, Mitochondrion intermembrane space Immunogen sequence: DGQYYGYLWS EVFSMDMFYS CFKKEGIMNP EVGMKYRNLI LKPGGSLDGM DMLHNFLKRE PNQKAFLMSR GLHAP.

This gene encodes a member of the metallopeptidase M3 protein family that cleaves neurotensin at the Pro10-Tyr11 bond, leading to the formation of neurotensin(1-10) and neurotensin(11-13). The encoded protein is likely involved in the termination of the neurotensinergic signal in the central nervous system and in the gastrointestinal tract.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLN
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Nln
Gene Accession No. P42676, Q9BYT8
Gene Alias 4930472G13Rik; AGTBP; angiotensin binding protein; angiotensin-binding protein; C79345; endopeptidase 24.16; EP24.16; KIAA1226; MEP; microsomal endopeptidase; Mitochondrial oligopeptidase M; MOP; neurolysin; neurolysin (metallopeptidase M3 family); neurolysin, mitochondrial; Neurotensin endopeptidase; NLN
Gene Symbols Nln
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 630-704 of human NLN (NP_0657771).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117041, 57486
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.