Learn More
NLN Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305580AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Neurolysin (EC 3.4.24.16) is an enzyme involved in the metabolic inactivation of bioactive peptides (Garrido et al., 1999 [PubMed 10235115]).Specifications
NLN | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9BYT8 | |
NLN | |
Between 25 and 75 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
NLN | |
AGTBP, EP24.16, MEP, MOP, angiotensin binding protein, angiotensin-binding protein, endopeptidase 24.16, microsomal endopeptidase, mitochondrial oligopeptidase M, neurolysin, mitochondrial, neurotensin endopeptidase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57486 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.