Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NLN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 147631AP150UL

Description
Neurolysin (EC 3.4.24.16) is an enzyme involved in the metabolic inactivation of bioactive peptides.Specifications
NLN | |
Polyclonal | |
Unconjugated | |
NLN | |
AGTBP, Angiotensin binding protein, KIAA1226, MEP, Microsomal endopeptidase, Mitochondrial oligopeptidase M, Neurolysin, mitochondrial, Neurotensin endopeptidase, NLN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117041, 57486, 75805 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P42676, Q91YP2, Q9BYT8 | |
NLN | |
NLN Fusion Protein Ag6524 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction