Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NLRC3 Antibody (Eowyn-1), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280084AF647
This item is not returnable.
View return policy
Description
NLRC3 Monoclonal antibody specifically detects NLRC3 in Mouse samples. It is validated for Western BlotSpecifications
NLRC3 | |
Monoclonal | |
Alexa Fluor 647 | |
CARD15-Like Protein, Caterpiller Protein 16.2, CLR16.2, Leucine Rich Repeat And CARD Domain Containing 3, NLR Family, CARD Domain Containing 3, NOD3, NOD-Like Receptor C3, Nucleotide-Binding Oligomerization Domain Protein 3, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And CARD Domain Containing 3, Protein NLRC3 | |
Recombinant mouse NLRC3 (card domain). | |
0.1 mL | |
Innate Immunity | |
197358 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot | |
Eowyn-1 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction