Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NLRC4 Monoclonal Antibody (GT1387)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527846

Catalog No. PIMA527846


Only null left
Add to Cart

Description

Description

Positive Control: human NLRC4-transfected 293T Predicted Reactivity: Bovine (82%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

NLRC4 encodes a member of the caspase recruitment domain-containing NLR family. Family members play essential roles in innate immune response to a wide range of pathogenic organisms, tissue damage and other cellular stresses. Mutations in this gene result in autoinflammation with infantile enterocolitis. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NLRC4
Monoclonal
1 mg/mL
PBS with no preservative
Q9NPP4
Nlrc4
Recombinant protein encompassing a sequence within the N-terminus region of human CARD12. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG2a
Western Blot, Immunocytochemistry
GT1387
Unconjugated
Nlrc4
9530011P19Rik; AIFEC; CARD, LRR, and NACHT-containing protein; Card12; caspase recruitment domain family, member 12; caspase recruitment domain-containing protein 12; CLAN; Clan protein; CLAN1; CLANA; CLANB; CLANC; CLAND; CLR2.1; FCAS4; ice protease-activating factor; ICE-protease activating factor; ipaf; NLR family CARD domain containing 4; NLR family CARD domain-containing protein 4; NLR family, CARD domain containing 4; NLRC4; NOD-like receptor C4; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 4; UNQ6189/PRO20215
Mouse
Protein A
RUO
58484
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.