Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nlrc4 (phospho S533) Rat anti-Human, Mouse, Clone:4B7B7, Abnova™
Rat monoclonal antibody raised against synthetic peptide of mouse Nlrc4 (phospho S533).
Supplier: Abnova Corporation MAB17177
Specifications
Nlrc4 (phospho S533) | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry Immunocytochemistry (1:200-1:1000) Flow Cytometry The optimal working dilution should be determined by the end user. |
|
Nlrc4 | |
Rat | |
100 μg | |
Human, Mouse | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Western Blot | |
4B7B7 | |
NLR family, CARD domain containing 4 | |
In PBS (0.05% sodium azide) | |
9530011P19Rik, CLAN, CLAN1, CLANA, CLANB, CLANC, CLAND, Card12, IPAF | |
A synthetic peptide corresponding to amino acid 525-538 of human Nlrc4 (phospho S533). | |
268973 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction