Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NLRC5 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$366.50 - $607.50
Specifications
Antigen | NLRC5 |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
NLRC5 Polyclonal antibody specifically detects NLRC5 in Human samples. It is validated for ImmunofluorescenceSpecifications
NLRC5 | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Immunology, Innate Immunity | |
PBS, pH 7.2, 40% glycerol | |
84166 | |
IgG | |
Affinity purified |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Caterpiller Protein 16.1, CLR16.1, NLR Family CARD Domain Containing 5, NOD27, NOD4, NOD-Like Receptor C5, Nucleotide-Binding Oligomerization Domain Protein 27, Nucleotide-Binding Oligomerization Domain Protein 4, Nucleotide-Binding Oligomerization Domains 27, Protein NLRC5 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: GLVRCFSTLQWLFRLDISFESQHILLRGDKTSRDMWATGSLPDFPAAAKFLGFRQRCIPRSLCLSECPLEPPSLTRLCATLKDCPGPLELQLSCEFLSDQ | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title