Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLRP1/NALP1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $316.50

Specifications

Antigen NLRP1/NALP1
Dilution Western Blot 1:1000 - 1:2000, Immunohistochemistry, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin 1:1000 - 1:5000
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056147T
SDP
View Documents
Novus Biologicals
NB10056147SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB10056147
SDP
View Documents
Novus Biologicals
NB10056147
0.05 mL
Each for $316.50
Only null left
Add to Cart
 
Description

Description

NLRP1/NALP1 Polyclonal specifically detects NLRP1/NALP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

NLRP1/NALP1
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Apoptosis
Whole antisera with 0.02% Sodium Azide
22861
A synthetic peptide corresponding to amino acids 1058-1077 (CVPSPASQGDLHTKPLGTDD) of human NALP1 (NAC/CARD7) was used as immunogen, GenBank no. NP_127497.1.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot 1:1000 - 1:2000, Immunohistochemistry, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin 1:1000 - 1:5000
Polyclonal
Antisera
RUO
Human
CARD7SLEV1, Caspase recruitment domain-containing protein 7, CLR17.1, Death effector filament-forming ced-4-like apoptosis protein, DEFCAPVAMAS1, DKFZp586O1822, KIAA0926DEFCAP-L/S, NACHT, leucine rich repeat and PYD (pyrin domain) containing 1, NACHT, leucine rich repeat and PYD containing 1, NACHT, LRR and PYD containing protein 1, NACHT, LRR and PYD domains-containing protein 1, NACPP1044, NALP1caspase recruitment domain protein 7, NLR family, pyrin domain containing 1, Nucleotide-binding domain and caspase recruitment domain, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 1
NLRP1
IgG
Unpurified
NALP1
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.