Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLRP2/NALP2 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056155T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056155T 0.025 mL
NB10056155 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056155T Supplier Novus Biologicals Supplier No. NB10056155SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

NLRP2/NALP2 Polyclonal specifically detects NLRP2/NALP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen NLRP2/NALP2
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:2000-1:5000
Formulation Whole antisera with 0.02% Sodium Azide
Gene Alias CLR19.9, FLJ20510, NACHT, leucine rich repeat and PYD containing 2, NACHT, LRR and PYD domains-containing protein 2, NALP2NACHT, LRR and PYD containing protein 2, NBS1PYRIN-containing APAF1-like protein 2, NLR family, pyrin domain containing 2, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 2, PAN1Nucleotide-binding site protein 1, PYPAF2PYRIN domain and NACHT domain-containing protein 1
Gene Symbols NLRP2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 97-115 (LKSFNKRKPLSLGITRKER) of human NALP2/PAN1/PYPAF2 was used as immunogen; GenBank no. gi 8923473 ref NP_060322.1
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 55655
Test Specificity Detected NALP2 at approx. 110 kDa in various cell line lysates and in NALP2 transfected cells (Bruey et al, 2004).
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.