Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NLRP3/NALP3 Antibody (Nalpy3-b) - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP191382
Description
Zinc finger protein pseudogene Polyclonal specifically detects Zinc finger protein pseudogene in Human samples. It is validated for Western Blot.Specifications
Zinc finger protein pseudogene | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
LOC728743 | |
Rabbit | |
22 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Zebra fish 83%. | |
Human, Mouse, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
LOC728743 | |
Synthetic peptide directed towards the C terminal of humanZinc finger protein pseudogene. Peptide sequence ARMPAPHPRRPGVFGERRPYFCPRCGKSFAREGSLKTHQRSHGHGPEGQA. | |
Affinity Purified | |
RUO | |
728743 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction