Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Anomalous homeobox protein Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | LOC647589 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Anomalous homeobox protein Polyclonal specifically detects Anomalous homeobox protein in Human samples. It is validated for Western Blot.Specifications
| LOC647589 | |
| Polyclonal | |
| Rabbit | |
| NP_001177983 | |
| 647589 | |
| Synthetic peptide directed towards the n terminal of human hCG_2007354. Peptide sequence LRLHLLDNADVALACARVLDQQEQQQAACRLLEGCQVPGGSQELVQLWND. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| anomalous homeobox, hypothetical protein LOC647589 | |
| ANHX | |
| IgG | |
| 42 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title