Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF90 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | ZNF90 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ZNF90 Polyclonal specifically detects ZNF90 in Human samples. It is validated for Western Blot.Specifications
ZNF90 | |
Polyclonal | |
Rabbit | |
Human | |
7643 | |
Synthetic peptide directed towards the N terminal of human ZNF90. Peptide sequence KDSFQKVIVTRYEKREYGNLELKKGCESVDEGKVHKRGYNGLNQCLTATQ. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
HTF9, zinc finger protein 90 | |
ZNF90 | |
IgG | |
72 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title