Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LKAAEAR1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$499.50
Specifications
Antigen | C20orf201 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
LKAAEAR1 Polyclonal specifically detects LKAAEAR1 in Human samples. It is validated for Western Blot.Specifications
C20orf201 | |
Polyclonal | |
Rabbit | |
NP_001007126 | |
C20orf201 | |
IgG | |
26 kDa |
Western Blot | |
Unconjugated | |
RUO | |
198437 | |
Synthetic peptide directed towards the C terminal of human LOC198437. Peptide sequence PGRGGERGGALPASAHAGPQPLSPAAEARGDHPGGERGWHHLPAVTATESV. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title