Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EXOC8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | EXOC8 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19144420
![]() |
Novus Biologicals
NBP19144420UL |
20 μL |
Each for $206.00
|
|
|||||
NBP191444
![]() |
Novus Biologicals
NBP191444 |
100 μL |
Each for $487.50
|
|
|||||
Description
EXOC8 Polyclonal specifically detects EXOC8 in Human samples. It is validated for Western Blot.Specifications
EXOC8 | |
Polyclonal | |
Rabbit | |
NP_787072 | |
149371 | |
Synthetic peptide directed towards the N terminal of human EXOC8. Peptide sequence MAMAMSDSGASRLRRQLESGGFEARLYVKQLSQQSDGDRDLQEHRQRIQA. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
exocyst complex component 8 | |
EXOC8 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title