Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MFSD4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | MFSD4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
MFSD4 Polyclonal specifically detects MFSD4 in Human samples. It is validated for Western Blot.Specifications
MFSD4 | |
Polyclonal | |
Rabbit | |
NP_857595 | |
148808 | |
Synthetic peptide directed towards the N terminal of human MFSD4. Peptide sequence FGALLSPLIADPFLSEANCLPANSTANTTSRGHLFHVSRVLGQHHVDAKP. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
DKFZp761N1114, FLJ25004, FLJ34577, FLJ45392, major facilitator superfamily domain containing 4, major facilitator superfamily domain-containing protein 4, UNQ3064 | |
MFSD4 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title