Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC144B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | CCDC144B |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CCDC144B Polyclonal specifically detects CCDC144B in Human samples. It is validated for Western Blot.Specifications
CCDC144B | |
Polyclonal | |
Rabbit | |
Human | |
284047 | |
Synthetic peptide directed towards the middle region of human CCDC144B. Peptide sequence TELSGTLTDGTTVGNDDDGLNQQIPRKENEEHDRPADKTANEKNKVKNQI. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
CCDC144B coiled-coil domain containing 144B (pseudogene) | |
CCDC144B | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title