Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC37A3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP191474 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP191474 100 μL
NBP19147420 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP191474 Supplier Novus Biologicals Supplier No. NBP191474
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRRC37A3 Polyclonal specifically detects LRRC37A3 in Human samples. It is validated for Western Blot.

Specifications

Antigen LRRC37A3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_955372
Gene Alias FLJ34306, KIAA0563, leucine rich repeat containing 37, member A3, leucine-rich repeat-containing protein 37A3, LRRC37A, MGC41826, MGC57168
Gene Symbols LRRC37A3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human LRRC37A3. Peptide sequence NYTSTELIIEPEEPSDSSGINLSGFGSEQLDTNDESDVTSTLSYILPYFS.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374819
Test Specificity Expected identity based on immunogen sequence: Human: 100%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.