Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p38 gamma/SAPK3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | p38 gamma/SAPK3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
p38 gamma/SAPK3 Polyclonal specifically detects p38 gamma/SAPK3 in Mouse samples. It is validated for Western Blot.Specifications
p38 gamma/SAPK3 | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication | |
EC 2.7.11, ERK3, ERK-6, ERK6MAPK 12, Extracellular signal-regulated kinase 6, MAP kinase 12, MAP kinase p38 gamma, mitogen-activated protein kinase 12, mitogen-activated protein kinase 3, Mitogen-activated protein kinase p38 gamma, P38GAMMA, PRKM12, SAPK-3, SAPK3EC 2.7.11.24, Stress-activated protein kinase 3 | |
MAPK12 | |
IgG | |
40 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_038899 | |
6300 | |
Synthetic peptide directed towards the C terminal of human Mapk12. Peptide sequence ERMLVLDAEQRVTAAEALTHPYFESLRDTEDEPKAQKYDDSFDDVDRTLE. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title