Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM83E Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | FAM83E |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
FAM83E Polyclonal specifically detects FAM83E in Human samples. It is validated for Western Blot.Specifications
| FAM83E | |
| Polyclonal | |
| Rabbit | |
| NP_060178 | |
| 54854 | |
| Synthetic peptide directed towards the middle region of human FAM83E. Peptide sequence RARTPSGPPARPSRSMWDLSRLSQLSGSSDGDNELKKSWGSKDTPAKALM. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| family with sequence similarity 83, member E, FLJ20200, hypothetical protein LOC54854, MGC138175, MGC138177 | |
| FAM83E | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title