Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRRM4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | SRRM4 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SRRM4 Polyclonal specifically detects SRRM4 in Human samples. It is validated for Western Blot.Specifications
| SRRM4 | |
| Polyclonal | |
| Rabbit | |
| NP_919262 | |
| 84530 | |
| Synthetic peptide directed towards the middle region of human SRRM4. Peptide sequence KTASPLTTSRGRSQEYDSGNDTSSPPSTQTSSARSRGQEKGSPSGGLSKS. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| KIAA1853, medulloblastoma antigen MU-MB-2.76, MU-MB-2.76, neural-specific SR-related protein of 100 kDa, nSR100, serine/arginine repetitive matrix 4, serine/arginine repetitive matrix protein 4 | |
| SRRM4 | |
| IgG | |
| 68 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title