Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFT2D3 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP191511

 View more versions of this product

Catalog No. NBP191511


Only null left
Add to Cart

Description

Description

SFT2D3 Polyclonal specifically detects SFT2D3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

SFT2D3
Polyclonal
Unconjugated
PBS, 2% Sucrose with 0.09% Sodium Azide
MGC5391, SFT2 domain containing 3, SFT2 domain-containing protein 3, vesicle transport protein SFT2C
Rabbit
22 kDa
100 μL
Primary
This product recognizes Human. Predicted Homology Based On Immunogen Sequence: yeast 72%.
Human, Yeast
IgG
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
NP_116129
SFT2D3
Synthetic peptide directed towards the N terminal of human SFT2D3. Peptide sequence EYLAQGKAGGPAAAEPLLAAEKAEEPGDRPAEEWLGRAGLRWTWARSPAE.
Affinity purified
RUO
84826
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.