Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKX2-4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | NKX2-4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1915420
![]() |
Novus Biologicals
NBP19154120UL |
20 μL |
Each for $206.00
|
|
|||||
NBP191541
![]() |
Novus Biologicals
NBP191541 |
100 μL |
Each for $487.50
|
|
|||||
Description
NKX2-4 Polyclonal specifically detects NKX2-4 in Human samples. It is validated for Western Blot.Specifications
NKX2-4 | |
Polyclonal | |
Rabbit | |
NP_075993 | |
644524 | |
Synthetic peptide directed towards the C terminal of mouse NKX2-4. Peptide sequence QQPQAPTPAPELEELSPSPPALHGPGGGLAALDAATGDYGGGVLGANLLY. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
homeobox protein Nkx-2.4, NK2 homeobox 4, NKX2.4, NKX2D | |
NKX2-4 | |
IgG | |
39 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title