Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C20orf203 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | C20orf203 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
C20orf203 Polyclonal specifically detects C20orf203 in Human samples. It is validated for Western Blot.Specifications
C20orf203 | |
Polyclonal | |
Rabbit | |
Human | |
alugen, chromosome 20 open reading frame 203, FLJ33706 | |
C20orf203 | |
IgG | |
221 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_872390 | |
284805 | |
Synthetic peptide directed towards the C terminal of human FLJ33706. Peptide sequence RMGTVGDFGQALSSLAWTSTCFQDFCLPSLPGKLPAPLISKQQFLSNSSR. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title