Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRWD1-IT2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | BRWD1-IT2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
BRWD1-IT2 Polyclonal specifically detects BRWD1-IT2 in Human samples. It is validated for Western Blot.Specifications
BRWD1-IT2 | |
Polyclonal | |
Rabbit | |
Human | |
257357 | |
Synthetic peptide directed towards the C terminal of human C21orf87. Peptide sequence LEARGLPCGARRTGPRRPVREMTLPSDPERATLPNPRLGAPAVPRRGPRS. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
BRWD1 intronic transcript 2 (non-protein coding), C21orf87, NCRNA00257 | |
BRWD1-IT2 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title